Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coronavirus Antibody (FIPV3-70), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064754PE
Description
Coronavirus Monoclonal specifically detects Coronavirus in Virus samples. It is validated for Flow Cytometry.Specifications
Coronavirus | |
Monoclonal | |
PE | |
Coronaviridae | |
Coronavirus cocktail | |
0.125 mL | |
Primary | |
Virus | |
Purified |
Flow Cytometry | |
FIPV3-70 | |
Flow Cytometry | |
Mouse | |
Protein A purified | |
RUO | |
NB100-64754 reacts with feline infectious peritonitis virus (FIPV) type 1 and 2 and is known to be specific for the nucleocapsid. It is also known to react with canine coronavirus (CCV), pig coronavirus transmissible gastroenteritis virus (TGEV) and ferret coronavirus. Some specific activity has been detected against bovine coronavirus (BCV). Clone FIPV3-70 exhibits negative reactivity against Feline Leukemia virus, Feline Immunodeficiency virus, Feline Calcivirus, Feline Herpes virus, Canine Adenovirus (type 2), Canine Distemper virus, Canine Parvovirus and Canine Parainfluenza virus. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction