Learn More
Anti-coronavirus (SARS-CoV-2) spike S1, mouse, mAb, MP Biomedicals™
Monoclonal antibody raised against the coronavirus spike S1 protein, a common target for neutralizing antibodies and vaccines.
$634.00
Specifications
Antigen | Coronavirus Spike S1 Protein |
---|---|
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Host Species | Mouse |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MP08720302
|
MP Biomedicals, Inc
08720302 |
250 μg |
Each of 1 for $634.00
|
|
|||||
Description
Anti-coronavirus (SARS-CoV-2) spike S1, mouse, mAb. Its genome encodes four structural proteins, i.e. spike (S), envelope (E), membrane (M), and nucleocapsid (N). The spike protein (S-protein) contains two subunits, S1 and S2. S1 defines the range of the hosts and specificity of the virus, thus recognizes and binds with the cell surface receptor. S2 subunit contains basic elements needed for the membrane fusion. The spike protein is the common target for neutralizing antibodies and vaccines.
Specifications
Coronavirus Spike S1 Protein | |
Monoclonal | |
Mouse | |
None | |
Primary |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
IgG1 |
For research or further manufacturing use only, not for direct human use.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.