Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anti-coronavirus (SARS-CoV-2) spike S2 (B), mouse, mAb, MP Biomedicals™
Monoclonal antibody raised against the coronavirus spike S2 (B) protein, a common target for neutralizing antibodies and vaccines.
$333.00
Specifications
| Antigen | Coronavirus Spike S2 (B) Protein |
|---|---|
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MP08720412
|
MP Biomedicals, Inc
08720412 |
250 μg |
Each of 1 for $333.00
|
|
|||||
Description
Anti-coronavirus (SARS-CoV-2) spike S2 (B), mouse, mAb. The 2019 Novel Coronavirus (SARS-CoV-2) is a coronavirus identified as the cause of an outbreak of respiratory illness first detected in Wuhan, China. Its genome encodes four structural proteins, i.e. spike (S), envelope (E), membrane (M) and nucleocapsid (N). The spike protein (S-protein) contains two subunits, S1 and S2. S1 defines the range of the hosts and specificity of the virus, thus recognizes and binds with the cell surface receptor. S2 subunit contains basic elements needed for the membrane fusion. The spike protein is the common target for neutralizing antibodies and vaccines.Specifications
| Coronavirus Spike S2 (B) Protein | |
| Monoclonal | |
| Mouse | |
| None | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| IgG1 |
For research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title