Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coronin-1a Antibody, Novus Biologicals™

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NB11058367
Description
Coronin-1a Polyclonal specifically detects Coronin-1a in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Coronin-1a | |
Polyclonal | |
Unconjugated | |
10mM Sodium PBS (0.9% isotonic, w/v, pH 7.2) with 0.02% Sodium Azide | |
CORO1A | |
Chickens were immunized with a synthetic peptide/keyhole limpet hemocyanin (KLH) conjugate. This synthetic peptide corresponded to a sequence shared between the human (NM_007074) and mouse (NM_009898) gene products. | |
0.5 mL | |
Cell Cycle and Replication, Immunology, Innate Immunity, Neuroscience | |
11151 | |
Human, Mouse, Non-species specific | |
IgY |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
Western Blot 1:5000-1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000-1:2000, Immunohistochemistry-Paraffin 1:10-1:500 | |
clipin-A, CLIPINA, CORO1, coronin, actin binding protein, 1A, coronin, actin-binding protein, 1A, coronin-1, coronin-1A, Coronin-like protein A, Coronin-like protein p57, FLJ41407, HCORO1, MGC117380, p57, TACOCLABP, Tryptophan aspartate-containing coat protein | |
Chicken | |
Affinity Purified | |
RUO | |
Primary | |
Coronin 1a | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction