Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Coronin 1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111327
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111327 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111327 Supplier Invitrogen™ Supplier No. PA5111327
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RELRVNRGLD TGRRRAAPEA SGTPSSDAVS RLEEEMRKLQ ATVQELQKRL DRLEETVQA Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 76%.

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Alternative splicing results in multiple transcript variants. A related pseudogene has been defined on chromosome 16.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coronin 1A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CORO1A
Gene Accession No. P31146
Gene Alias actin-binding; bovine p57; Clabp; Clabp coronin; CLIPINA; Clipin-A; CORO1; CORO1A; CORO1A protein; coronin 1; coronin 1A; coronin, actin binding protein 1A; coronin, actin binding protein, 1A; coronin-1; coronin-1A; coronin-like actin binding protein; coronin-like protein A; Coronin-like protein p57; FLJ41407; HCORO1; IMD8; Lmb3; MGC117380; p57; TACO; tryptophan aspartate-containing coat protein
Gene Symbols CORO1A
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Coronin 1A. Recombinant protein control fragment (Product #RP-100865).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11151
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.