Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Coronin 1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546001
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546001 Supplier Invitrogen™ Supplier No. PA546001
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: STTTAADATP SGSLARAGEA GKLEEVMQEL RALRALVKEQ GDRICRLEEQ Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 86%.

Members of the coronin family, such as CORO1B, are WD repeat-containing actin-binding proteins that regulate cell motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coronin 1B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Coro1b
Gene Accession No. Q9BR76
Gene Alias cor-1; Coro1b; coronin 1b; coronin 2; coronin, actin binding protein 1B; coronin, actin binding protein, 1B; coronin, actin binding protein, 1B (coronin 2); coronin, actin-binding protein, 1B; coronin-1B; coronin-1B-like; CORONIN-2; Coronin-like protein pp66; coroninse
Gene Symbols Coro1b
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CORO1B (aa 431-480).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57175
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.