Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Coronin 1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589926
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589926 Supplier Invitrogen™ Supplier No. PA589926
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSGRDADPIL ISLREAYVPS KQRDLKISRR NVLSDSRPAM APGSSHLGAP ASTTTAADAT PSGSLARAGE AGKLEEVMQE LRALRALVKE QGDRICRLEE QLGRMENGDA; Positive Samples: Rat kidney; Cellular Location: Cytoplasm, cytoskeleton.

Members of the coronin family, such as CORO1B, are WD repeat-containing actin-binding proteins that regulate cell motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coronin 1B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Coro1b
Gene Accession No. O89046
Gene Alias cor-1; Coro1b; coronin 1b; coronin 2; coronin, actin binding protein 1B; coronin, actin binding protein, 1B; coronin, actin binding protein, 1B (coronin 2); coronin, actin-binding protein, 1B; coronin-1B; coronin-1B-like; CORONIN-2; Coronin-like protein pp66; coroninse
Gene Symbols Coro1b
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 380-489 of human CORO1B (NP_0651741).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29474
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.