Learn More
Invitrogen™ Coronin 1B Polyclonal Antibody

Description
Recommended positive controls: 293T, Molt-4, Raji, NIH-3T3. Predicted reactivity: Mouse (98%), Rat (98%), Rabbit (96%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Coronin 1B |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Coro1b |
| Gene Accession No. | Q9BR76, Q9WUM3 |
| Gene Alias | cor-1; Coro1b; coronin 1b; coronin 2; coronin, actin binding protein 1B; coronin, actin binding protein, 1B; coronin, actin binding protein, 1B (coronin 2); coronin, actin-binding protein, 1B; coronin-1B; coronin-1B-like; CORONIN-2; Coronin-like protein pp66; coroninse |
| Gene Symbols | Coro1b |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.