Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Coronin 2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559458
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559458 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559458 Supplier Invitrogen™ Supplier No. PA559458
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DIYPPTAGAQ PSLTAQEWLS GMNRDPILVS LRPGSELLRP HPLPAERPIF NSMAPASPRL LNQTEKLAAE DGW Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 66%.

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein contains 5 WD repeats, and has a structural similarity with actin-binding proteins: the D. discoideum coronin and the human p57 protein, suggesting that this protein may also be an actin-binding protein that regulates cell motility. Alternative splicing of this gene generates 2 transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coronin 2A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CORO2A
Gene Accession No. Q92828
Gene Alias CLIPINB; CORO2A; coronin 2A; coronin, actin binding protein, 2A; coronin, actin-binding protein, 2A; coronin-2A; coronin-like protein B; IR10; WD protein IR10; WD repeat-containing protein 2; WDR2; WD-repeat protein 2
Gene Symbols CORO2A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Coronin 2A. Recombinant protein control fragment (Product #RP-98381).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7464
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.