Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Coronin 2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569819
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
May play a role in the reorganization of neuronal actin structure.
Specifications
Coronin 2B | |
Polyclonal | |
Unconjugated | |
CORO2B | |
clipin C; CLIPINC; Clipin-C; CORO2B; coronin 2B; coronin, actin binding protein, 2B; coronin-2B; Coronin-like protein C; E130012P22; E130012P22Rik; KIAA0925; Protein FC96 | |
Rabbit | |
Affinity chromatography | |
RUO | |
235431 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8BH44 | |
CORO2B | |
synthetic peptide directed towards the N-terminal region of mouse Coronin 2B. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction