Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coronin 2B Polyclonal Antibody

Coronin 2B Polyclonal Antibody
Supplier: Thermo Scientific PA569820
Description
Coronin 2B Polyclonal Antibody for Western Blot

Specifications
Coronin 2B | |
Polyclonal | |
Unconjugated | |
CORO2B | |
clipin C; CLIPINC; Clipin-C; CORO2B; coronin 2B; coronin, actin binding protein, 2B; coronin-2B; Coronin-like protein C; E130012P22; E130012P22Rik; KIAA0925; Protein FC96 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
235431 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8BH44 | |
CORO2B | |
synthetic peptide corresponding to a region of mouse Coronin 2B (aa 275-324). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction