Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coronin 2B Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572048
Description
CORO2B may play a role in the reorganization of neuronal actin structure.
Specifications
Coronin 2B | |
Polyclonal | |
Unconjugated | |
CORO2B | |
Coronin-2B; Coronin-like protein C; Clipin-C; Protein FC96; CORO2B; KIAA0925 | |
Rabbit | |
Protein A | |
RUO | |
10391 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q9UQ03 | |
CORO2B | |
KLH conjugated synthetic peptide between 1-27 amino acids from the N-terminal region of human CORO2B | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction