Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Coronin 6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524758
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524758 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524758 Supplier Invitrogen™ Supplier No. PA524758
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with rat and rabbit based on sequence homology.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Coronin 6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CORO6
Gene Accession No. Q6QEF8, Q920M5
Gene Alias clipin E; clipin-E; Coro6; coronin 6; coronin 6/clipin E type D; coronin relative protein; coronin, actin binding protein 6; coronin-6; Coronin-like protein E; PP1009; PP1782; PP1881
Gene Symbols CORO6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 337-365 amino acids from the C-terminal region of human CORO6.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216961, 84940
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.