Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Coronin 7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572695
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572695 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572695 Supplier Invitrogen™ Supplier No. PA572695
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Coronin 7 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Coronins, such as CORO7, constitute an evolutionarily conserved family of WD-repeat actin-binding proteins. CORO7 plays a role in Golgi complex morphology and function (Rybakin et al., 2004, 2006 [PubMed 15327992] [PubMed 16905771]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coronin 7
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CORO7
Gene Accession No. O35828, P57737, Q9D2V7
Gene Alias 0610011B16Rik; 70 kDa WD repeat tumor rejection antigen; 70 kDa WD repeat tumor rejection antigen homolog; AW048373; AW556392; CORO7; coronin 7; coronin-7; CRN7; POD1; tumor specific antigen 70 kDa
Gene Symbols CORO7
Host Species Rabbit
Immunogen Coronin 7 antibody for an 17 amino acid synthetic peptide near the carboxy terminus of human Coronin 7. The immunogen is within the last 50 amino acids of Coronin 7.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192276, 78885, 79585
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.