Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cortactin Antibody (4C6), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237621
Description
Cortactin Monoclonal specifically detects Cortactin in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cortactin | |
Monoclonal | |
Unconjugated | |
Ascites with 0.03% Sodium Azide | |
CTTN | |
Purified recombinant fragment of human CTTN expressed in E. Coli. | |
Unpurified | |
RUO | |
Primary | |
Human, Mouse | |
Ascites |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
4C6 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
1110020L01Rik, Amplaxin, cortactin, ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 srcsubstrate), EMS1amplaxin, FLJ34459, Oncogene EMS1, src substrate cortactin | |
Mouse | |
80 kDa | |
0.1 mL | |
Cytoskeleton Markers | |
2017 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction