Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corticotropin Releasing Factor Antibody (4H9), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP321462
Description
Corticotropin Releasing Factor Monoclonal antibody specifically detects Corticotropin Releasing Factor in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
Corticotropin Releasing Factor | |
Monoclonal | |
1 mg/mL | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
corticoliberin, corticotropin releasing hormone, Corticotropin-releasing hormone, CRFCorticotropin-releasing factor | |
Corticotrophin Releasing Factor conjugated to Bovine Serum Albumin | |
0.2 mg | |
Cell Biology, Endocrinology, Neuroscience, Signal Transduction | |
1392 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
4H9 | |
Unconjugated | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction