Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corticotropin Releasing Factor Antibody - Azide and BSA Free, Novus Biologicals™

Sheep Polyclonal Antibody has been used in 7 publications
Supplier: Novus Biologicals NB11081721
Description
Corticotropin Releasing Factor Polyclonal specifically detects Corticotropin Releasing Factor in Human, Mouse, Rat, Guinea Pig (Negative) samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Corticotropin Releasing Factor | |
Polyclonal | |
Unconjugated | |
0.05X PBS with No Preservative | |
CRH | |
An 18 amino acid synthetic CRF peptide (H-CEQLAQQAHSNRKLMEII-OH), conjugated to Keyhole Limpet Hemocyanin (KLH). | |
0.02 mg | |
Primary | |
Specific for Corticotropin Releasing Factor. Confirmed by Immunohistochemical analysis of Paraffin Embedded Rat Brain (Hypothalamus). | |
Human, Mouse, Rat, Guinea Pig (Negative) | |
IgG |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
LYOPH | |
ELISA 1:100-1:10, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:10-1:500 | |
corticoliberin, corticotropin releasing hormone, Corticotropin-releasing hormone, CRFCorticotropin-releasing factor | |
Sheep | |
Affinity Purified | |
RUO | |
1392 | |
Reconstitute with 0.02 ml sterilized water. | |
Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction