Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corticotropin Releasing Factor Monoclonal Antibody (5D5), QED Bioscience™
SDP

Catalog No. 502278146
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8146 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8146 Supplier Leinco Technologies Inc Supplier No. 56102100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Corticotropin Releasing Factor Monoclonal antibody specifically detects Corticotropin Releasing Factor in Human, Rat samples. It is validated for Inhibition Assays

Specifications

Antigen Corticotropin Releasing Factor
Applications Inhibition Assays
Classification Monoclonal
Clone 5D5
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene CRH
Gene Accession No. P01143, P06850
Gene Alias Corticoliberin; corticotropin releasing factor; corticotropin releasing hormone; corticotropin releasing hormone receptor 2; corticotropin-releasing factor; Corticotropin-releasing factor receptor 2; corticotropin-releasing hormone; Corticotropin-releasing hormone receptor 2; CRF; CRF2; CRF2 receptor, beta isoform; CRF2R; CRFR2; CRF-R-2; CRF-RB; CRH; CRH receptor 2 variant B; CRH1; CRH2R; CRHR2; CRH-R2; CRH-R-2; Gm1347; HM-CRF
Host Species Mouse
Immunogen Purified human/rat corticotropin-releasing factor (CRF, Sigma).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1392, 81648
Target Species Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.