Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cortisol Antibody (BGN/C53), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062484AF750
Description
Cortisol Monoclonal specifically detects Cortisol in All Species samples. It is validated for ELISA.Specifications
Cortisol | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
Synthetic corticosterone conjugated to Bovine Serum Albumin. | |
Primary | |
All species | |
IgG3 |
ELISA | |
BGN/C53 | |
ELISA | |
Mouse | |
0.1 mL | |
This product recognises the glucocorticoid hormone cortisol, produced in the adrenal cortex and involved in responses to stress. Cross reactivity: Cortisol 100%, Corticosterone 50%, Prednisolone 15%, 11-Desoxycortisol, <0.1% Cortisone,<0.1% 17-a-OH Progesterone, <0.1%. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction