Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cortisol Monoclonal Antibody (F4P1A3)
GREENER_CHOICE

Catalog No. PIMIC0201
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMIC0201 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMIC0201 Supplier Invitrogen™ Supplier No. MIC0201
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MIC0201 targets Cortisol in ELISA, IP, and RIA applications and shows reactivity with Chemical samples. The MIC0201 immunogen is cortisol-3-CMO-BSA. MIC0201 was formerly sold as a Seradyn product.

Cortisol is the most potent glucocorticoid produced by the human adrenal. It is synthesized from cholesterol and its production is stimulated by pituitary adrenocorticotropic hormone (ACTH) which is regulated by corticotropin releasing factor (CRF). ACTH and CRF secretions are inhibited by high cortisol levels in a negative feedback loop. In plasma a majority of cortisol is bound with high affinity to corticosteroid binding globulin (CBG or transcotin). Cortisol acts through specific intracellular receptors and affects numerous physiologic systems including immune function, glucose counter regulation, vascular tone, and bone metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cortisol
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone F4P1A3
Concentration 5.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Cortisol-3-CMO-BSA.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.