Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Cortisol Parameter Assay Kit
SDP

Catalog No. SKGE008B Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SKGE008B 1 Pk. (6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SKGE008B Supplier R&D Systems™ Supplier No. SKGE008B

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cortisol Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Cortisol Parameter Assay Kit quantifies multi-species Cortisol in cell culture supernates (10 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), saliva (20 ul), urine (10 ul).

Specifications

Product Type ELISA Kit
Quantity 1 Pk. (6 Plates)
Species Multi-species
Conjugate Biotin
Binding Target Cortisol
Assay Sensitivity 0.111 ng/mL
Assay Range 0.16 to 10 ng/mL
For Use With (Application) ELISA
Product Line R&D Systems
Sample Type Cell Culture Supernates (10uL), Serum (10uL), EDTA Plasma (10uL), Heparin Plasma (10uL), Saliva (20uL), Urine (10uL)
Test Time 2.5 hr.
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Endocrinology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.