Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cortisone, 98.1%, MP Biomedicals™

Catalog No. icn19017201 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 53-06-5
Molecular Formula C21H28O5
Molecular Weight (g/mol) 360.45
MDL Number MFCD00003610
InChI Key MFYSYFVPBJMHGN-ZPOLXVRWSA-N
Synonym cortisone, andreson, cortisal, cortisate, cortistal, cortivite, cortogen, cortone, kendall's compound e, adrenalex
PubChem CID 222786
ChEBI CHEBI:16962
IUPAC Name (1R,3aS,3bS,9aR,9bS,11aS)-1-hydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-7,10-dione
SMILES C[C@]12CC(=O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO

Specifications

Color White
Assay Percent Range 0.981
Quantity 1 g
Formula Weight 360.4
Physical Form Powder
Chemical Name or Material Cortisone
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.