Learn More
AquaPhoenix Scientific Corundum
SCI_ED
Catalog No. S02886 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Product Type:
Mineral
Catalog No. Product Type
S02886 Mineral
1 options
Catalog No. S02886 Supplier AquaPhoenix Scientific Supplier No. EM5303B
Add to Cart
Add to Cart
This 1 to 1.25 inches corundum specimen is an excellent example of one of the hardest minerals on Earth—perfect for classroom use and hardness testing.
  • Natural corundum mineral specimen
  • Size range: approximately 1 to 1.25 in.
  • Second hardest mineral on the Mohs scale (hardness = 9)
  • Typically shows a dull luster and hexagonal crystal habit
  • Ideal for mineral identification and hardness comparison
  • Great for geology labs, rock and mineral collections, or educational kits
  • Suitable for middle school through college-level earth science
Specifications
Product Type Mineral