Learn More
Eisco™ Corundum Mineral Specimen
SCI_ED
Catalog No. S24668 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Speciman
12/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S24668 12/Pk.
S24667 1 Speciman
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S24668 Supplier Eisco™ Supplier No. ESNG0008PK12
Add to Cart
Add to Cart
For study of mineral hardness and crystal habit - EISCO Corundum Mineral Specimen is used in classroom identification labs and exercises, correlates well to Next Generation Science Standards Evidence Statements.
  • For exercises designed to distinguish between crystal habit and cleavage
  • Comes in clear labeled and barcoded bag for ideal storage
  • Average specimen size: 2-3cm
  • Has Mohs scale hardness of 9, making it one of the hardest minerals on planet
  • For use in mineral hardness exercises and lessons in differentiating between cleavage and crystal habit
Specifications
Product Type Mineral
For Use With (Application) Study of mineral hardness and crystal habit
Packaging Clear labeled and barcoded bag
Quantity 12/Pk.
Specimen Size Approx. 1 in.
Specimen Type Corundum