Corning™ 96-Well Clear Bottom Black or White Polystyrene Microplates Available on GSA/VA Contract for Federal Government customers only.

Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Clear Bottom Black or White Polystyrene Microplates permit direct microscopic viewing. For fluorescent and luminescent assays; ideal for performing cell-based and molecular assays.

Manufacturer: Corning™ 3603

 View more versions of this product

Catalog No. 07-200-565

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    For Use With (Application) Attachment and growth of anchorage-dependent cells
    Binding Property TC-treated
    Bottom Flat bottom
    Lid Yes
    Material Polystyrene
    Sterile Yes
    Type 96-Well Assay Microplate
    Volume (Metric) Well 360μL
    Volume (Metric) Working 75 to 200μL
    Closure Yes
    Color Black
    Applications Fluorescent and luminescent assays, performing cell-based and molecular assays
    Surface Coating Tissue culture treated

    • Molded polystyrene with opaque walls and optically clear bottoms
    • Bottoms are 60% thinner than conventional polystyrene microplates for lower background flourecence and enabling readings down to 340nm
    • Opaque black or white walls prevent well to well crosstalk
    • Recommended working volumes of 75 to 200µL
    • Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
    • Treated surface options are not coated, require no special handling and are stable at room temperature
      • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
      • Ideal for serial dilutions, hemagglutination or precipitation assays, and general storage applications
      • Plate dimensions meet proposed industry standards

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit