Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Costar™ 96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate
BIOPHARMAGSA_VAGREENER_CHOICE

Catalog No. p-9045050
Click to view available options
Packaging:
Bag of 20; Cs. of 100
Individually wrapped, 50/Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

Color: Clear; Sterility: Sterile; Lid: With Lid

Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.

  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or nonsterile

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid With Lid
Culture Area 0.32 cm2
Array 8 x 12
Packaging Individually wrapped, 50/Cs.
Binding Type TC-Treated
Color Clear
Product Type Cell Culture Microplate
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment Cell Culture
Total Volume (Metric) Well 0.33 mL
Well Shape Round Bottom (U-well)
For Use With (Application) Attachment and growth of anchorage-dependent cells
No. per Case 50/Cs.
No. per Pack 1/Pk.
Quantity 50/Cs.
Well Color Clear
Diameter (Metric) Well 0.64 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.