Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ 96-Well Clear PVC Assay Microplates
Shop All Corning Products
Click to view available options
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round
Description
Order Info
Lids are not available. Supplied in packs of 25.
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Color | Clear |
| Material | PVC |
| No. of Wells | 96 |
| Volume (Metric) Working | 50 to 150 μL |
| Sterility | Non-sterile |
| Surface Treatment | Non-treated |
| Well Shape | Conical Bottom (V-well) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction