Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Costar™ Transwell™-COL Collagen-Coated PTFE Membrane Insert

Catalog No. 07200153 Shop All Corning Products
Click to view available options
Pore Size:
0.4 μm
3 μm
For Use With (Equipment):
12 Well Plate
24 Well Plate
6 Well Plate
This item is not returnable. View return policy

Catalog No. 07-200-153

Supplier: Corning™ 3495

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Transparent, collagen-treated microporous membranes promote cell attachment and spreading

Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.

You can use cell culture inserts to:

  • Produce a cell culture environment that closely resembles an in vivo state
  • Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings Transparent, Collagen-coated PTFE membrane
  • Promotes cell attachment and spreading
  • Equimolar mixture of Collagen type I and III
  • Individually packaged
  • Multiple well plates included in each case
  • Sterile

Order Info

Individually packed and supplied in package with tissue culture plates with lids; sterile.

TRUSTED_SUSTAINABILITY

Specifications

Culture Area 0.33 cm2
Material PTFE
Volume (Metric) Well 0.6 mL
Pore Size 0.4 μm
Type Membrane Insert
For Use With (Equipment) 24 Well Plate
Diameter (Metric) Outer 6.5 mm
No. per Pack 1/Pk.
No. per Case 24/Cs.
Surface Coating Collagen
Sterility Sterile
No. of Wells 24
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.