Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COTL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119036
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119036 200 μL
PIPA5122072 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119036 Supplier Invitrogen™ Supplier No. PA5119036
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

COTL1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes one of the numerous actin-binding proteins which regulate the actin cytoskeleton. This protein binds F-actin, and also interacts with 5-lipoxygenase, which is the first committed enzyme in leukotriene biosynthesis. Although this gene has been reported to map to chromosome 17 in the Smith-Magenis syndrome region, the best alignments for this gene are to chromosome 16. The Smith-Magenis syndrome region is the site of two related pseudogenes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COTL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Cotl1
Gene Accession No. Q14019, Q9CQI6
Gene Alias 1810074P22Rik; 2010004C08Rik; CLP; coactosin like F-actin binding protein 1; coactosin-like 1; coactosin-like 1 (Dictyostelium); coactosin-like F-actin binding protein 1; coactosin-like protein; coactosin-like protein {ECO:0000250; Cotl1; cotl1 {ECO:0000312; EMBL:AAI58747.1, ECO:0000312; FLJ43657; MGC137989 protein; MGC19733; RGD:1305498}; UniProtKB:Q14019}; Unknown (protein for MGC:63983); wu:fb08h08; zgc:63983
Gene Symbols Cotl1
Host Species Rabbit
Immunogen Recombinant protein Coactosin Like Protein 1. The antigen corresponds to amino acid range 1-142 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23406, 72042
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.