Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cotton Rat CCL2/JE/MCP-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | CCL2/JE/MCP-1 |
---|---|
Clone | 238607 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Cotton Rat samples. It is validated for Western Blot.Specifications
CCL2/JE/MCP-1 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Cotton Rat | |
AAN85636 | |
6347 | |
E. coli-derived recombinant cotton rat CCL2/JE/MCP-1 Gln25-Asn150 Accession # AAN85636 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects cotton rat CCL2/JE/MCP-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse CCL2 is observed and no cross-reactivity with recombinant human CCL1, 2, 3, 5, 7, 8, 11, 13, 14, 15, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, recombinant mouse CCL1, 3, 4, 5, 6, 7, 9, 11, 12, 17, 19, 20, 21, 22, 24, 25, or recombinant rat CCL20 is observed. |
238607 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF | |
CCL2 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title