Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cotton Rat CCL3/MIP-1 alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1061SP
This item is not returnable.
View return policy
Description
CCL3/MIP-1 alpha Monoclonal specifically detects CCL3/MIP-1 alpha in Cotton Rat samples. It is validated for Western Blot, Neutralization.Specifications
CCL3/MIP-1 alpha | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Neutralization 0.05-0.3 ug/mL | |
AAL26704 | |
CCL3 | |
E. coli-derived recombinant cotton rat CCL3/MIP-1 alpha Ala24-Ala92 Accession # AAL26704 | |
25 μg | |
Primary | |
Detects cotton rat CCL3/MIP-1 alpha in direct ELISAs. In direct ELISAs, this antibody shows approximately 50% cross-reactivity with recombinant human CCL3/MIP-1 alpha and recombinant cotton rat CCL4/MIP-1 beta and no cross-reactivity with recombinant cotton rat CCL5, recombinant human CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, 27, 28, recombinant mouse CCL1, 2, 3, 4, 5, 6, 7, 8, 9/10/MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, 27, 28, or recombinant rat CCL20. | |
Cotton Rat | |
Purified |
Western Blot, Neutralization | |
175024 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6348 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction