Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cotton Rat CCL4/MIP-1 beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1024SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1024SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1024SP Supplier R&D Systems Supplier No. AF1024SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CCL4/MIP-1 beta Polyclonal specifically detects CCL4/MIP-1 beta in Cotton Rat samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CCL4/MIP-1 beta
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 0.15-0.6 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAL16933
Gene Alias ACT2, ACT-2, AT744.1, C-C motif chemokine 4, chemokine (C-C motif) ligand 4, Exodus-3, G-26, G-26 T-lymphocyte-secreted protein, HC21, LAG1, LAG-1, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, MIP1 beta, MIP-1 beta, MIP1B, MIP1B1, MIP-1-beta, MIP-1-beta(1-69), PAT 744, Protein H400, SCYA2, SCYA4, secreted protein G-26, SIS-gamma, small inducible cytokine A4 (homologous to mouse Mip-1b), Small-inducible cytokine A4, T-cell activation protein 2
Gene Symbols CCL4
Host Species Goat
Immunogen E. coli-derived recombinant cotton rat CCL4/MIP-1 beta Ala24-Asn92 Accession # AAL16933
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6351
Test Specificity Detects cotton rat CCL4/MIP-1 beta in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human MIP-1 beta is observed and less than 2% cross-reactivity with recombinant mouse MIP-1 beta is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Cotton Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.