Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cotton Rat CD8 alpha alpha Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB7080G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7080G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7080G Supplier R&D Systems Supplier No. FAB7080G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD8 Monoclonal specifically detects CD8 in Cotton Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen CD8
Applications Flow Cytometry
Classification Monoclonal
Clone JG12
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. AAL55392
Gene Alias CD8, CD8A, CD8a molecule, LEU2, p32
Gene Symbols CD8A
Host Species Mouse
Immunogen NIH-3T3 mouse embryonic fibroblast cell line transfected with cotton rat CD8 alpha and SP2/O mouse melanoma cell line transfected with cotton rat CD8 alpha . Accession # AAL55392
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 925
Test Specificity Detects cotton rat CD8 alpha in direct ELISAs.
Target Species Cotton Rat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.