Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cotton Rat CD8 alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7080
This item is not returnable.
View return policy
Description
CD8 Monoclonal specifically detects CD8 in Cotton Rat samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
CD8 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
AAL55392 | |
CD8A | |
NIH-3T3 mouse embryonic fibroblast cell line transfected with cotton rat CD8 alpha and SP2/O mouse melanoma cell line transfected with cotton rat CD8 alpha . Accession # AAL55392 | |
100 μg | |
Primary | |
Detects cotton rat CD8 alpha in direct ELISAs. | |
Cotton Rat | |
Purified |
Flow Cytometry, CyTOF | |
JG12 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD8, CD8A, CD8a molecule, LEU2, p32 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
925 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction