Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cotton Rat CXCL1/2/3/GRO Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1114SP
This item is not returnable.
View return policy
Description
CXCL1/2/3/GRO Monoclonal specifically detects CXCL1/2/3/GRO in Cotton Rat samples. It is validated for Western Blot, Neutralization.Specifications
CXCL1/2/3/GRO | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Neutralization 0.08-0.4 ug/mL | |
AAL16934 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2919 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Neutralization | |
186413 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1 | |
E. coli-derived recombinant cotton rat GRO Ala28-Lys100 Accession # AAL16934 | |
25 μg | |
Primary | |
Detects cotton rat CXCL1/2/3/GRO in direct ELISAs and Western blots. In direct ELISAs, this antibody shows 5-20% cross-reactivity with recombinant rat CXCL1 and recombinant human CXCL2 and no cross-reactivity with recombinant cotton rat CXCL10, recombinant human CXCL1, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12/SDF-1 alpha , rh12/SDF-1 beta , 13, 16, recombinant mouse CXCL1, 2, 4, 6, 7, 9, 10, 11, 12/SDF-1 alpha , 13, 15, 16, 17, recombinant rat (rr) CXCL2, rrCXCL3/CINC-2 alpha , rr3/CINC-2 beta , or recombinant porcine CXCL8. | |
Cotton Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction