Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cotton Rat CXCL1/2/3/GRO Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1114
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1114 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1114 Supplier R&D Systems Supplier No. MAB1114
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL1/2/3/GRO Monoclonal specifically detects CXCL1/2/3/GRO in Cotton Rat samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CXCL1/2/3/GRO
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 186413
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.08-0.4 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAL16934
Gene Alias FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1
Host Species Mouse
Immunogen E. coli-derived recombinant cotton rat GRO Ala28-Lys100 Accession # AAL16934
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2919
Test Specificity Detects cotton rat CXCL1/2/3/GRO in direct ELISAs and Western blots. In direct ELISAs, this antibody shows 5-20% cross-reactivity with recombinant rat CXCL1 and recombinant human CXCL2 and no cross-reactivity with recombinant cotton rat CXCL10, recombinant human CXCL1, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12/SDF-1 alpha , rh12/SDF-1 beta , 13, 16, recombinant mouse CXCL1, 2, 4, 6, 7, 9, 10, 11, 12/SDF-1 alpha , 13, 15, 16, 17, recombinant rat (rr) CXCL2, rrCXCL3/CINC-2 alpha , rr3/CINC-2 beta , or recombinant porcine CXCL8.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Cotton Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.