Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cotton Rat CXCL10/IP-10/CRG-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1117
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1117 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1117 Supplier R&D Systems Supplier No. MAB1117
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL10/IP-10/CRG-2 Monoclonal specifically detects CXCL10/IP-10/CRG-2 in Cotton Rat samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CXCL10/IP-10/CRG-2
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 163426
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 20-40 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAL16935
Gene Alias C7, chemokine (C-X-C motif) ligand 10, crg-2, CRG2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10
Gene Symbols CXCL10
Host Species Mouse
Immunogen E. coli-derived recombinant cotton rat CXCL10/IP-10/CRG-2 Ile22-Val94 Accession # AAL16935
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3627
Test Specificity Detects cotton rat CXCL10/IP-10/CRG-2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CXCL1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 13, or 16, recombinant mouse CXCL1, 2, 4, 6, 7, 9, 10, 11, 12, 13, 15 or 16, recombinant rat CXCL1, 2, 3, 5 or recombinant porcine CXCL8 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Cotton Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.