Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cotton Rat IFN-alpha Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1082
This item is not returnable.
View return policy
Description
IFN-alpha Polyclonal specifically detects IFN-alpha in Cotton Rat samples. It is validated for Western Blot.Specifications
IFN-alpha | |
Polyclonal | |
Biotin | |
AAL18816 | |
Goat | |
Affinity Purified | |
RUO | |
3440 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D | |
E. coli-derived recombinant cotton rat IFN-alpha (R&D Systems, Catalog # 1082-CR) Cys24-Lys190 with a Cys109Tyr substitution Accession # AAL18816 | |
50 μg | |
Primary | |
Detects cotton rat IFN-alpha in Western blots. In this format, less than 1% cross-reactivity with recombinant human IFN-beta and recombinant cotton rat IFN-gamma is observed. | |
Cotton Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction