Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cotton Rat IFN-gamma Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF565
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF565 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF565 Supplier R&D Systems Supplier No. BAF565
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 4 publications

IFN-gamma Polyclonal specifically detects IFN-gamma in Cotton Rat samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen IFN-gamma
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. Q9QXX2
Gene Alias IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Gene Symbols IFNG
Host Species Goat
Immunogen E. coli-derived recombinant cotton rat IFN-gamma His21-Ile170 Accession # Q9QXX2
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3458
Test Specificity Detects cotton rat IFN-gamma in ELISAs and Western blots. In sandwich immunoassays, less than 0.3% cross-reactivity with recombinant human (rh) IFN-gamma , recombinant mouse (rm) IFN-gamma , recombinant rat (rr) IFN-gamma , recombinant porcine (rp) IFN-gamma , recombinant canine IFN-gamma , and recombinant feline (rfe) IFN-gamma is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Cotton Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.