Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cotton Rat IL-1 alpha/IL-1F1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF552
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF552 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF552 Supplier R&D Systems Supplier No. AF552
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-1 alpha/IL-1F1 Polyclonal specifically detects IL-1 alpha/IL-1F1 in Cotton Rat samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen IL-1 alpha/IL-1F1
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 0.015-0.045 ug/mL
Gene Alias BAF, Hematopoietin-1, IL1, IL1 alpha, IL-1 alpha, IL1A, IL-1A, IL1-ALPHA, IL1F1, IL-1F1, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, LAF, LEM, preinterleukin 1 alpha, pro-interleukin-1-alpha
Gene Symbols IL1A
Host Species Goat
Immunogen E. coli-derived recombinant cotton rat IL-1 alpha/IL-1F1
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3552
Test Specificity Detects cotton rat IL-1 alpha/IL-1F1 in direct ELISAs and Western blots. In Western blots, approximately 40% cross-reactivity with recombinant rat IL-1 alpha is observed, 15% cross-reactivity with recombinant mouse IL-1 alpha is observed, and less than 1% cross-reactivity with recombinant human IL-1 alpha and recombinant porcine IL-1 alpha is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Cotton Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.