Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cotton Rat IL-1 beta/IL-1F2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1009SP
This item is not returnable.
View return policy
Description
IL-1 beta/IL-1F2 Polyclonal specifically detects IL-1 beta/IL-1F2 in Cotton Rat samples. It is validated for Western Blot, Neutralization.Specifications
IL-1 beta/IL-1F2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IL1B | |
E. coli-derived recombinant cotton rat IL-1 beta | |
25 μg | |
Primary | |
Detects cotton rat IL-1 beta/IL-1F2 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human IL-1 beta and recombinant mouse IL-1 beta is observed, and 10% cross-reactivity with recombinant rat IL-1 beta and recombinant porcine IL-1 beta is observed. | |
Cotton Rat | |
IgG |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.4-1.2 ug/mL | |
catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
Goat | |
Antigen Affinity-purified | |
RUO | |
3553 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction