Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cotton Rat TNF-alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB10111
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Cotton Rat samples. It is validated for Western Blot, Neutralization.Specifications
| TNF-alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 0.04-0.2 ug/mL | |
| AAL18818 | |
| TNF | |
| E. coli-derived recombinant cotton rat TNF-alpha Leu1-Leu156 Accession # AAL18818 | |
| 500 μg | |
| Primary | |
| Detects cotton rat TNF-alpha in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) TNF-alpha is observed and no cross-reactivity with rhAPRIL, rhBAFF, rhEDA-A2, recombinant mouse (rm) EDA, rhFas Ligand, rhGITR Ligand, rhLIGHT, rhOX40 Ligand, rmTNF-alpha , recombinant porcine TNF-alpha , recombinant rat TNF-alpha , recombinant rhesus macaque TNF-alpha , rhTRAIL, rmTRAIL, rhTRANCE, rhTWEAK, or rhVEGI is observed. | |
| Cotton Rat | |
| Purified |
| Western Blot, Neutralization | |
| 159813 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7124 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction