Learn More
Coulomb's Law Apparatus
SDPSCI_ED
Catalog No. s52054
Click to view available options
:
Coulombs Law Apparatus
Includes
  • Stable base
  • Draft shield
  • Support on which to suspend filaments
  • Viewing window
  • Measuring scales
  • Selection of electrostatic materials
Experiment with electrical forces and charges.
Used in PSSC experiments dealing with electrical forces and charges.
Specifications
For Use With (Application) Explore the force between charged particles by varying the charge and the distance
Includes Stable base, draft shield, support on which to suspend filaments, viewing window, measuring scales, selection of electrostatic materials
Product Type Coulombs Law Apparatus
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.