Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coumarin 314, 99%, pure, laser grade

Catalog No. AC405620025
Change view
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC405620025 2.5 g
AC405621000 100 mg
AC405625000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC405620025 Supplier Thermo Scientific Chemicals Supplier No. 405620025
Add to Cart

CAS: 55804-66-5 | C18H19NO4 | 313.353 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55804-66-5
Molecular Formula C18H19NO4
Molecular Weight (g/mol) 313.353
MDL Number MFCD00051334
InChI Key VMJKUPWQKZFFCX-UHFFFAOYSA-N
Synonym 1,2,4,5,3H,6H,10H-Tetrahydro-9-carbethoxy[1]benzopyrano(9,9a,1-gh)-quinolizin-10-one
PubChem CID 72653
ChEBI CHEBI:51940
SMILES CCOC(=O)C1=CC2=CC3=C4C(=C2OC1=O)CCCN4CCC3

Specifications

Melting Point 140.0°C to 144.0°C
Assay Percent Range 99%
Formula Weight 313.34
Percent Purity 99%
Grade Pure
Quantity 2.5 g
Chemical Name or Material Coumarin 314

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.