Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coumarin 6, 98%, pure

Catalog No. p-94009
Click to view available options
Quantity:
500 mg
2.5 g

CAS: 38215-36-0 | C20H18N2O2S | 350.436 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38215-36-0
Molecular Formula C20H18N2O2S
Molecular Weight (g/mol) 350.436
MDL Number MFCD00041869
InChI Key VBVAVBCYMYWNOU-UHFFFAOYSA-N
Synonym 3-(2'-Benzothiazolyl)-7-diethylaminocoumarin
PubChem CID 100334
ChEBI CHEBI:51942
IUPAC Name 3-(1,3-benzothiazol-2-yl)-7-(diethylamino)chromen-2-one
SMILES CCN(CC)C1=CC2=C(C=C1)C=C(C(=O)O2)C3=NC4=CC=CC=C4S3

Specifications

Assay Percent Range 98%
Fluorescence Max 497nm (in methanol)
Infrared Spectrum Authentic
Lambda Max 458nm (in methanol)
Formula Weight 350.44
Grade Extra Pure
Molar Absorption Coefficient 40800L/(mole.cm) (lambda max.)
Packaging Glass bottle
Suitability (for use as a laser dye) passes
UV_Vis 97.5% min.
Color Orange
Quantity 500 mg
Physical Form Powder
Chemical Name or Material Coumarin 6, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 253-830-2
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.