Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8a Antibody [RPA-T8] (PerCP-Cy5.5), ProSci
SDP

Catalog No. 16832556
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-832-556 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-832-556 Supplier ProSci LLC Supplier No. 76710T025
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD8a Monoclonal Antibody specifically detects CD8a in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CD8a
Applications Flow Cytometry
Classification Monoclonal
Clone RPA-T8
Conjugate PerCP-Cyanine5.5
Formulation Phosphate-buffered aqueous solution, ≤0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
Gene Alias CD8, MAL, p32, Leu2, CD8A
Host Species Mouse
Purification Method The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 925
Test Specificity The RPA-T8 monoclonal antibody specifically reacts with the human CD8a molecule, a 32 kDa cell surface receptor expressed either as a heterodimer (CD8 alpha/beta) or as a homodimer (CD8 alpha/alpha) on the majority of thymocytes, a subpopulation of mature T cells, and natural killer cells.
Target Species Human
Content And Storage The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Research Category Immunology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.