Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COUP-TF I/NR2F1 Antibody (SR1792), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | COUP-TF I/NR2F1 |
---|---|
Clone | SR1792 |
Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
COUP-TF I/NR2F1 Monoclonal antibody specifically detects COUP-TF I/NR2F1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
COUP-TF I/NR2F1 | |
Western Blot 1:500-1:1000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
7025 | |
IgG | |
Affinity purified |
SR1792 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
COUP transcription factor 1, COUP-TF1, COUP-TFI, EAR-3COUP transcription factor I, EAR3COUP-TF I, ERBAL3V-erbA-related protein 3, Nuclear receptor subfamily 2 group F member 1, nuclear receptor subfamily 2, group F, member 1, SVP44, TCFCOUP1, TFCOUP1NR2F2, transcription factor COUP 1 (chicken ovalbumin upstream promoter 1, v-erb-ahomolog-like 3) | |
A synthesized peptide derived from human COUP-TF I/NR2F1 (Uniprot #: P10589) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title