Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COUP-TF II/NR2F2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP131980 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP131980 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP131980 Supplier Novus Biologicals Supplier No. NBP131980
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COUP-TF II/NR2F2 Polyclonal specifically detects COUP-TF II/NR2F2 in Mouse, Porcine samples. It is validated for Western Blot, Knockdown Validated.

Specifications

Antigen COUP-TF II/NR2F2
Applications Western Blot, KnockDown
Classification Polyclonal
Concentration 0.57mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:20000, KnockDown Validated
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias Apolipoprotein A-I regulatory protein 1, ARP-1, ARP1ADP-ribosylation factor related protein 1, chicken ovalbumin upstream promoter transcription factor 2, COUP transcription factor 2, COUP transcription factor II, COUP-TF II, COUP-TF2, COUPTFB, COUPTFII, COUP-TFII, MGC117452, Nuclear receptor subfamily 2 group F member 2, nuclear receptor subfamily 2, group F, member 2, SVP40, TFCOUP2apolipoprotein AI regulatory protein 1
Gene Symbols NR2F2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NR2F2. The exact sequence is proprietary.
Molecular Weight of Antigen 46 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 7026
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Mouse, Pig
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.