Learn More
Bollé Safety Covarsi Safety Goggles
Covarsi safety goggles maximum safety for all including wearers of prescription glasses, polycarbonate, OTG, features high wrap around
Supplier: Bollé Safety COVERSI
Description
- Economical and effective
- Comfortable, flexible frame
- Adjustable strap with pivoting buckle
Specifications
Anti-fog, Scratch-resistant | |
Rectangular | |
Safety Goggles | |
3.42 oz. | |
Polycarbonate | |
Sealed |
ANSI Z87+ | |
Clear | |
Clear | |
Reusable | |
UVA and UVB |