Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Covidien Dermacea™ 100% Cotton Gauze Sponges

Catalog No. p-4265972
Click to view available options
Dimensions (L x W):
2 x 2 in.
3 x 3 in.
4 x 4 in.
Thickness:
12-Ply
8-Ply
Sterility:
Non-sterile
Sterile
This item is not returnable. View return policy
This item is not returnable. View return policy

An alternative for sterile and nonsterile 100% cotton gauze dressings

Specifications

Material Cotton
Product Type Gauze Sponge
Sterility Sterile
Quantity 3000 Cs.
Dimensions (L x W) 2 x 2 in.
For Use With (Application) Wound dressing, general cleaning, minor prepping, wound packing and debriding
Thickness 8-Ply
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.